Learn More
Med Vet International AM Induction Chamber - Small (1.2L), J0265P, Each
Catalog No. 5000495859
Encompass_Preferred
Tiny size for mice, small exotics/birds. ? Sealed lid w/gasket and two side clips ? Fresh gas inlet is metal barbed fitting with 5? of smooth vinyl tubing w/proximal 15mm ET connector ? Proximal end has 15mm o.d. connector ? Outlet Gas ? 22mm corrugated tubing Dimensions: 4"W x 6"L x 3"H
Catalog No. 50-004-95859 Supplier Med Vet International Supplier No. JOR265P
Tiny size for mice, small exotics/birds. ? Sealed lid w/gasket and two side clips ? Fresh gas inlet is metal barbed fitting with 5? of smooth vinyl tubing w/proximal 15mm ET connector ? Proximal end has 15mm o.d. connector ? Outlet Gas ? 22mm corrugated tubing Dimensions: 4"W x 6"L x 3"H