Learn More
Andwin Scientific CHAMBER CAP 1ML 200/CS
SDP
Catalog No. NC9875089
Encompass_Preferred
The 1 mL chamber cap is an accessory for the Cyto-Tek Cyto Centrifuge, which is an instrument made to move cells from suspension onto a glass slide by the process of cytocentrifugation. The equipment can hold up to 12 specimen holders. The chamber caps are sold in a case of 200.
Catalog No. NC9875089 Supplier Andwin Scientific Supplier No. 4335
The 1 mL chamber cap is an accessory for the Cyto-Tek Cyto Centrifuge, which is an instrument made to move cells from suspension onto a glass slide by the process of cytocentrifugation. The equipment can hold up to 12 specimen holders. The chamber caps are sold in a case of 200.