Learn More
Darwin Chambers Company Environmental Chamber 29.75""W x 23""D x 27.75""H.
SDP
Catalog No. 501251776
Encompass_Preferred
The KB011 is a 11.6 cubic feet bench top environmental chamber. Temperature range is 2 to 50 degrees C. Humidity range ambient to 90 percent. This unit comes with 2 stainless steel shelves. The chamber provides 304 series stainless steel interior and door liner, corrosion-resistant; also a heavy-gauge powder-coated, galvanized steel exterior. The KB series chambers feature hot gas by-pass technology which controls the temperature of the epoxy coated evaporator for greater uniformity. The ultrasonic humidification system provides excellent humidity control and avoids hot spots seen during chamber mapping of steam boiler equipped chambers.
Catalog No. 50-1251776 Supplier Darwin Chambers Company Supplier No. KB011
Add to Cart
Add to Cart
Request a Quote
The KB011 is a 11.6 cubic feet bench top environmental chamber. Temperature range is 2 to 50 degrees C. Humidity range ambient to 90 percent. This unit comes with 2 stainless steel shelves. The chamber provides 304 series stainless steel interior and door liner, corrosion-resistant; also a heavy-gauge powder-coated, galvanized steel exterior. The KB series chambers feature hot gas by-pass technology which controls the temperature of the epoxy coated evaporator for greater uniformity. The ultrasonic humidification system provides excellent humidity control and avoids hot spots seen during chamber mapping of steam boiler equipped chambers.