Learn More
Darwin Chambers Company INSECT CHAMBER 24 CUFT
SDP
Catalog No. 501251761
Encompass_Preferred
The IN030 is a 23.9 cubic foot single door insect rearing chamber. Temp range is 18 to 60 deg C. Optional humidity range ambient to 90%. Humidity and lighting options not included. Must be ordered separately. This unit comes w/ 3 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Silver-colored, polyester-coated aluminum interior & stainless steel floor w/ coved corners. The 7 year warranty on cooling components & the ability of our chambers to maintain temp. even if the chamber suffers multiple failures is unmatched by any other refrigeration-based chamber. The ultrasonic humidification system provides excellent humidity control & avoids hot spots seen during chamber mapping of steam boiler equipped chambers. The IN Series chambers utilize less energy than comparable insect rearing chambers.
Catalog No. 50-1251761 Supplier Darwin Chambers Company Supplier No. IN030AA
Add to Cart
Add to Cart
Request a Quote
The IN030 is a 23.9 cubic foot single door insect rearing chamber. Temp range is 18 to 60 deg C. Optional humidity range ambient to 90%. Humidity and lighting options not included. Must be ordered separately. This unit comes w/ 3 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Silver-colored, polyester-coated aluminum interior & stainless steel floor w/ coved corners. The 7 year warranty on cooling components & the ability of our chambers to maintain temp. even if the chamber suffers multiple failures is unmatched by any other refrigeration-based chamber. The ultrasonic humidification system provides excellent humidity control & avoids hot spots seen during chamber mapping of steam boiler equipped chambers. The IN Series chambers utilize less energy than comparable insect rearing chambers.