Learn More
Darwin Chambers Company CHAMBER INSECT 30 CU. FOOT
SDP
Catalog No. 501251763
Encompass_Preferred
The IN034 is 32.1 cubic feet a extra wide single door insect rearing chamber. Temperature range is 16°C to 50°C. Optional humidity range ambient to 90 percent. Humidity and lighting options not included. Must be ordered separately. This unit comes with 3 stainless steel shelves. The chamber provides 304 series stainless steel interior and door liner, corrosion-resistant; also a heavy-gauge powder-coated, galvanized steel exterior. The 7 year warranty on cooling components and the ability of our chambers to maintain temperature even if the chamber suffers multiple failures is unmatched by any other refrigeration-based chamber. The ultrasonic humidification system provides excellent humidity control and avoids hot spots seen during chamber mapping of steam boiler equipped chambers. The IN Series chambers utilize less energy than comparable insect rearing chambers.
Catalog No. 50-1251763 Supplier Darwin Chambers Company Supplier No. IN034
Add to Cart
Add to Cart
Request a Quote
The IN034 is 32.1 cubic feet a extra wide single door insect rearing chamber. Temperature range is 16°C to 50°C. Optional humidity range ambient to 90 percent. Humidity and lighting options not included. Must be ordered separately. This unit comes with 3 stainless steel shelves. The chamber provides 304 series stainless steel interior and door liner, corrosion-resistant; also a heavy-gauge powder-coated, galvanized steel exterior. The 7 year warranty on cooling components and the ability of our chambers to maintain temperature even if the chamber suffers multiple failures is unmatched by any other refrigeration-based chamber. The ultrasonic humidification system provides excellent humidity control and avoids hot spots seen during chamber mapping of steam boiler equipped chambers. The IN Series chambers utilize less energy than comparable insect rearing chambers.