Learn More
Darwin Chambers Company CHAMBER INSECT 70 CU FOOT
SDP
Catalog No. 501251766
Encompass_Preferred
The IN084 is a 76.2 cubic foot triple door insect rearing chamber. Temp range is 18 to 60 deg C. Optional humidity range ambient to 90%. Humidity and lighting options not included. Must be ordered separately. This unit comes w/ 9 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Silver-colored, polyester-coated aluminum interior & stainless steel floor w/ coved corners. The 7 year warranty on cooling components & the ability of our chambers to maintain temp. even if the chamber suffers multiple failures is unmatched by any other refrigeration-based chamber. The ultrasonic humidification system provides excellent humidity control & avoids hot spots seen during chamber mapping of steam boiler equipped chambers. The IN Series chambers utilize less energy than comparable insect rearing chambers.
Catalog No. 50-1251766 Supplier Darwin Chambers Company Supplier No. IN084AA
Add to Cart
Add to Cart
Request a Quote
The IN084 is a 76.2 cubic foot triple door insect rearing chamber. Temp range is 18 to 60 deg C. Optional humidity range ambient to 90%. Humidity and lighting options not included. Must be ordered separately. This unit comes w/ 9 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Silver-colored, polyester-coated aluminum interior & stainless steel floor w/ coved corners. The 7 year warranty on cooling components & the ability of our chambers to maintain temp. even if the chamber suffers multiple failures is unmatched by any other refrigeration-based chamber. The ultrasonic humidification system provides excellent humidity control & avoids hot spots seen during chamber mapping of steam boiler equipped chambers. The IN Series chambers utilize less energy than comparable insect rearing chambers.