Learn More
Darwin Chambers Company Bench Top Insect rearing chamber 29.75""W x 23""D x 27.75""H
SDP
Catalog No. 501251768
Encompass_Preferred
The IN011 is 11.6 cubic feet a benchtop insect rearing chamber. Temperature range is 12°C to 50°C. Optional humidity range ambient to 90 percent. Humidity and lighting options not included. Must be ordered separately. This unit comes with 2 stainless steel shelves. The chamber provides 304 series stainless steel interior and door liner, corrosion-resistant; also a heavy-gauge powder-coated, galvanized steel exterior. The 7 year warranty on cooling components and the ability of our chambers to maintain temperature even if the chamber suffers multiple failures is unmatched by any other refrigeration-based chamber. The ultrasonic humidification system provides excellent humidity control and avoids hot spots seen during chamber mapping of steam boiler equipped chambers. The IN Series chambers utilize less energy than comparable insect rearing chambers.
Catalog No. 50-1251768 Supplier Darwin Chambers Company Supplier No. IN011
Add to Cart
Add to Cart
Request a Quote
The IN011 is 11.6 cubic feet a benchtop insect rearing chamber. Temperature range is 12°C to 50°C. Optional humidity range ambient to 90 percent. Humidity and lighting options not included. Must be ordered separately. This unit comes with 2 stainless steel shelves. The chamber provides 304 series stainless steel interior and door liner, corrosion-resistant; also a heavy-gauge powder-coated, galvanized steel exterior. The 7 year warranty on cooling components and the ability of our chambers to maintain temperature even if the chamber suffers multiple failures is unmatched by any other refrigeration-based chamber. The ultrasonic humidification system provides excellent humidity control and avoids hot spots seen during chamber mapping of steam boiler equipped chambers. The IN Series chambers utilize less energy than comparable insect rearing chambers.