Learn More
Electron Microscopy Sciences Anodized Aluminum Microbead Chamber, Red
SDP
Catalog No. 501927581
Encompass_Preferred
Use with Thermal Microbeads to create a one size fits all dry bath solution. Fits all standard sized dry bath systems. Microbeads tightly surround tubes, microtubes, and bottles to evenly distribute temperatures. Single and double place chambers allow maximum flexibility when working with dry baths. Use with dry ice in our popular pans and buckets for full customized sample cooling.
Color: Red
Catalog No. 50-192-7581 Supplier Electron Microscopy Sciences Supplier No. 6860530
Use with Thermal Microbeads to create a one size fits all dry bath solution. Fits all standard sized dry bath systems. Microbeads tightly surround tubes, microtubes, and bottles to evenly distribute temperatures. Single and double place chambers allow maximum flexibility when working with dry baths. Use with dry ice in our popular pans and buckets for full customized sample cooling.
Color: Red