Learn More
Beckman Coulter O-Rings
Catalog No. NC0442277
Encompass
Beckman Coulter offers a wide selection of O-rings in various sizes and materials for use in laboratory equipment and instrumentation. O-rings are available in Buna-N, Viton, EPR, Silicone, PTFE, Aflas, and other materials. Sizes range from 1mm ID to 50mm ID. O-rings can be purchased individually or in packs. Beckman Coulter O-rings meet ASTM D1418 standards and are manufactured for optimal performance and durability in laboratory environments. O-rings can be used for fluid sealing, vibration dampening, and impact cushioning in analytical instruments, centrifuges, and other devices.
Catalog No. NC0442277 Supplier Beckman Coulter Supplier No. 801778
Beckman Coulter offers a wide selection of O-rings in various sizes and materials for use in laboratory equipment and instrumentation. O-rings are available in Buna-N, Viton, EPR, Silicone, PTFE, Aflas, and other materials. Sizes range from 1mm ID to 50mm ID. O-rings can be purchased individually or in packs. Beckman Coulter O-rings meet ASTM D1418 standards and are manufactured for optimal performance and durability in laboratory environments. O-rings can be used for fluid sealing, vibration dampening, and impact cushioning in analytical instruments, centrifuges, and other devices.