Learn More
Electron Microscopy Sciences Double Neubauer Ruled Hemacytometer Chamber Only (3510)
SDP
Catalog No. 5031291
Encompass_Preferred
Double improved Neubauer rulings are diamond etched directly into a polished recess to reduce damage from scratches. Opaque surfaces above and below the counting area offer an easy focal reference. The chamber comes with a quick orientation mark for the microscope. 0.1 mm cell depth.
Catalog No. 50-312-91 Supplier Electron Microscopy Sciences Supplier No. 6351113
Double improved Neubauer rulings are diamond etched directly into a polished recess to reduce damage from scratches. Opaque surfaces above and below the counting area offer an easy focal reference. The chamber comes with a quick orientation mark for the microscope. 0.1 mm cell depth.