Learn More
Chemglass Life Sciences Chamber Slide, 1 Well, White, with Glass Slide, Sterile (6/pk, 2pks/Case)
SDP
Catalog No. 501897784
Encompass_Preferred
Chamber Cell Culture Slide from Nest allows scientists to grow cells directly on a microscope slide without need for further transfer. The microscope slide is a standard size, 25 mm x 75 mm. NEST cell culture chambered slides are designed for various cell-based experiments and are also useful for virus, toxicity tests and immunocytological experiments. Each chamber slide is composed of a base made of polypropylene, the slide which is made of borosilicate glass, the chambers which are made of polystyrene. The slides come in packs of 6 slides/per tray and the tray can be used as an incubation rack in your CO2 incubator.
Catalog No. 50-189-7784 Supplier Chemglass Life Sciences Supplier No. CGN3530001W
Chamber Cell Culture Slide from Nest allows scientists to grow cells directly on a microscope slide without need for further transfer. The microscope slide is a standard size, 25 mm x 75 mm. NEST cell culture chambered slides are designed for various cell-based experiments and are also useful for virus, toxicity tests and immunocytological experiments. Each chamber slide is composed of a base made of polypropylene, the slide which is made of borosilicate glass, the chambers which are made of polystyrene. The slides come in packs of 6 slides/per tray and the tray can be used as an incubation rack in your CO2 incubator.