Learn More
Perkin Elmer US LLC Quartz Baffled Cyclonic Spray Chamber for ELAN DRC/DRCplus/DRC II
Catalog No. 509046878
Encompass_Preferred
The spray chamber is an integral part of the sample introduction system responsible for ltering the sample mist to permit the appropriate droplet size distribution to reach the plasma. The baffed cyclonic spray chamber features a central transfer tube which acts as a secondary particle separator helping separate larger aerosol particles from the sample. This reduces solvent load in the plasma without compromising detection limits. The distance between the bottom of the baffle and the chamber wall is carefully controlled so that droplets cannot form on the bottom of the baffle. The advantage is smooth draining and improved precision.
Catalog No. 50-904-6878 Supplier Perkin Elmer US LLC Supplier No. WE025221
The spray chamber is an integral part of the sample introduction system responsible for ltering the sample mist to permit the appropriate droplet size distribution to reach the plasma. The baffed cyclonic spray chamber features a central transfer tube which acts as a secondary particle separator helping separate larger aerosol particles from the sample. This reduces solvent load in the plasma without compromising detection limits. The distance between the bottom of the baffle and the chamber wall is carefully controlled so that droplets cannot form on the bottom of the baffle. The advantage is smooth draining and improved precision.