Learn More
Darwin Chambers Company Stability Chamber 29.75""W x 23.125""D x 29.75""H.
SDP
Catalog No. 501251786
Encompass_Preferred
The PH011 is a 11.6 cubic ft benchtop stability chamber that can be stacked w/ optional racking. These Chambers are designed for performance specifications exceeding FDA & ICH requirements. Temperature range is 12°C to 50°C degrees C. Humidity Range ambient to 90 percent. This unit comes w/ two stainless steel shelves. The chamber interior provides a 304 series stainless steel interior & door liner, corrosion-resistant; also heavy-gauge powder-coated, galvanized steel exterior. The 7 year warranty on cooling components & the ability of our chambers to maintain temperature even if the chamber suffers multiple failures is unmatched by any other refrigeration-based chamber. The ultrasonic humidification system provides excellent humidity control & avoids hot spots seen during chamber mapping of steam boiler equipped chambers. The PH Series chambers utilize less energy than comparable stability chambers.
Catalog No. 50-1251786 Supplier Darwin Chambers Company Supplier No. PH011
Add to Cart
Add to Cart
Request a Quote
The PH011 is a 11.6 cubic ft benchtop stability chamber that can be stacked w/ optional racking. These Chambers are designed for performance specifications exceeding FDA & ICH requirements. Temperature range is 12°C to 50°C degrees C. Humidity Range ambient to 90 percent. This unit comes w/ two stainless steel shelves. The chamber interior provides a 304 series stainless steel interior & door liner, corrosion-resistant; also heavy-gauge powder-coated, galvanized steel exterior. The 7 year warranty on cooling components & the ability of our chambers to maintain temperature even if the chamber suffers multiple failures is unmatched by any other refrigeration-based chamber. The ultrasonic humidification system provides excellent humidity control & avoids hot spots seen during chamber mapping of steam boiler equipped chambers. The PH Series chambers utilize less energy than comparable stability chambers.