Learn More
Perkin Elmer US LLC UltraClean PFA PTFE Scott Spray Chamber for Optima 2x00/4x00/5x00/7x00 DV/8x00
Catalog No. 509045583
Encompass_Preferred
This spray chamber is for use with concentric nebulizers. It provides high aerosol transmission and excellent short/long-term stability. Constructed from PFA for low contamination and long lifetime, the O-ring-free seal reduces contamination and provides an easy-to-clean, low-maintenance system. Applications include semiconductor, high precision, and hydrofluoric acid samples. Features: PFA PTFE construction Low contamination Chemically inert Ideal for dilute-and-shoot analysis O-ring-free seal reduces contamination Easy to clean, low maintenance High transmission and selectivity
Catalog No. 50-904-5583 Supplier Perkin Elmer US LLC Supplier No. N8122356
Add to Cart
Add to Cart
Request a Quote
This spray chamber is for use with concentric nebulizers. It provides high aerosol transmission and excellent short/long-term stability. Constructed from PFA for low contamination and long lifetime, the O-ring-free seal reduces contamination and provides an easy-to-clean, low-maintenance system. Applications include semiconductor, high precision, and hydrofluoric acid samples. Features: PFA PTFE construction Low contamination Chemically inert Ideal for dilute-and-shoot analysis O-ring-free seal reduces contamination Easy to clean, low maintenance High transmission and selectivity