Learn More
Electron Microscopy Sciences CultureWell™ Chambered Coverglass 12 wells, 15 mm dia, 250-400 mcL
SDP
Catalog No. 50930305
Encompass_Preferred
The thin flat coverglass is the optimal platform for high resolution microscopy and high- sensitivity, quantitative, fluorescent imaging. Leak-proof design eliminates well-to-well cross contamination. The chambered coverglass products consists of removable / reusable non-cytotoxic silicone gaskets secured to 24 mm x 50 mm, #1.5 German coverglass. They may be placed in sterile culture plates or 100 mm diameter culture dishes.
Volume: 250-400 µl
I.D.: 15 mm dia
Depth: 1 mm
Catalog No. 50-930-305 Supplier Electron Microscopy Sciences Supplier No. 704602R1
The thin flat coverglass is the optimal platform for high resolution microscopy and high- sensitivity, quantitative, fluorescent imaging. Leak-proof design eliminates well-to-well cross contamination. The chambered coverglass products consists of removable / reusable non-cytotoxic silicone gaskets secured to 24 mm x 50 mm, #1.5 German coverglass. They may be placed in sterile culture plates or 100 mm diameter culture dishes.
Volume: 250-400 µl
I.D.: 15 mm dia
Depth: 1 mm