Learn More
Electron Microscopy Sciences CultureWell™ Chambered Coverglass 13 wells, 9.5x9.5mm, 300-500 mcL
SDP
Catalog No. 50930307
Encompass_Preferred
The thin flat coverglass is the optimal platform for high resolution microscopy and high- sensitivity, quantitative, fluorescent imaging. Leak-proof design eliminates well-to-well cross contamination. The chambered coverglass products consists of removable / reusable non-cytotoxic silicone gaskets secured to 24 mm x 50 mm, #1.5 German coverglass. They may be placed in sterile culture plates or 100 mm diameter culture dishes.
Volume: 300-500 µl
I.D.: 9.5 x 9.5 mm
Depth: 1 mm
Catalog No. 50-930-307 Supplier Electron Microscopy Sciences Supplier No. 704603S
The thin flat coverglass is the optimal platform for high resolution microscopy and high- sensitivity, quantitative, fluorescent imaging. Leak-proof design eliminates well-to-well cross contamination. The chambered coverglass products consists of removable / reusable non-cytotoxic silicone gaskets secured to 24 mm x 50 mm, #1.5 German coverglass. They may be placed in sterile culture plates or 100 mm diameter culture dishes.
Volume: 300-500 µl
I.D.: 9.5 x 9.5 mm
Depth: 1 mm