Learn More
Electron Microscopy Sciences CultureWell™ Chambered Coverglass 18 wells, 6mm Dia, 15-30 mcL
SDP
Catalog No. 50930310
Encompass_Preferred
The thin flat coverglass is the optimal platform for high resolution microscopy and high- sensitivity, quantitative, fluorescent imaging. Leak-proof design eliminates well-to-well cross contamination. The chambered coverglass products consists of removable / reusable non-cytotoxic silicone gaskets secured to 24 mm x 50 mm, #1.5 German coverglass. They may be placed in sterile culture plates or 100 mm diameter culture dishes.
Volume: 15-30 µl
I.D.: 6 mm Dia
Depth: 1 mm
Catalog No. 50-930-310 Supplier Electron Microscopy Sciences Supplier No. 704608R
The thin flat coverglass is the optimal platform for high resolution microscopy and high- sensitivity, quantitative, fluorescent imaging. Leak-proof design eliminates well-to-well cross contamination. The chambered coverglass products consists of removable / reusable non-cytotoxic silicone gaskets secured to 24 mm x 50 mm, #1.5 German coverglass. They may be placed in sterile culture plates or 100 mm diameter culture dishes.
Volume: 15-30 µl
I.D.: 6 mm Dia
Depth: 1 mm