Learn More
Electron Microscopy Sciences CultureWell™ Coverglass Inserts 8x4 wells, 300-500 mcL, 15 mm I.D., 1.5 mm Depth
SDP
Catalog No. 50930312
Encompass_Preferred
Each CultureWell insert is comprised of four chambered coverglass assembled in a disposable frame which is placed in a standard 86 mm x 128 mm culture plate.  All are sterile and ready to use. The stabilizing insert design and accessory hardware allow fragile coverglass to be easily integrated into protocols for high throughput handling of microscope slides and SBS compliant microtiter plates.
Volume: 300-500 µl
I.D.: 15 mm
Catalog No. 50-930-312 Supplier Electron Microscopy Sciences Supplier No. 704612R2
Each CultureWell insert is comprised of four chambered coverglass assembled in a disposable frame which is placed in a standard 86 mm x 128 mm culture plate.  All are sterile and ready to use. The stabilizing insert design and accessory hardware allow fragile coverglass to be easily integrated into protocols for high throughput handling of microscope slides and SBS compliant microtiter plates.
Volume: 300-500 µl
I.D.: 15 mm