Learn More
World Precision Instrument Chambers for Ussing System-CHM1
SDP
Supplier:  World Precision Instrument CHM1
Encompass
WPI’s classical Ussing Chambers are well established perfusion chambers that are easy to operate, easy to control temperature, and easy to clean after use. The Ussing Chambers are machined from solid acrylic with eight entry ports for fluid lines, electrodes, or agar bridges. For easy, leak-free attachment of tubing and electrodes, all eight ports are luer type. The four ports for voltage and current electrodes are recessed to prevent formation of air bubbles in the chamber. The fluid compartments in each side of the chamber are separated by the epithelial membrane being studied. Sharp stainless steel pins on one side of the chamber hold the membrane in position and mate with holes in the opposite chamber interface.
Catalog No. 50-253-8899