Learn More
LICORbio Chameleon 700 Pre-stained Protein Ladder (500 L)
Catalog No. NC0769375
Encompass_Preferred
The Chameleon 700 Pre-stained Protein Ladder offers multi-colored, pre-stained proteins for both visible and 700 channel near-infrared detection. It includes 6 protein bands from 8 to 160 kDa. Use for easy visualization of gel migration and protein size, and to orient your gel or membrane quickly.
Catalog No. NC0769375 Supplier LICORbio Supplier No. 92870000
The Chameleon 700 Pre-stained Protein Ladder offers multi-colored, pre-stained proteins for both visible and 700 channel near-infrared detection. It includes 6 protein bands from 8 to 160 kDa. Use for easy visualization of gel migration and protein size, and to orient your gel or membrane quickly.