Learn More
LICORbio IRDye 800CW DBCO Infrared Dye (0.5 mg)
Catalog No. NC0769376
Encompass_Preferred
The Chameleon 800 Pre-stained Protein Ladder offers multi-colored, pre-stained proteins for both visible and 800 channel near-infrared detection. It includes 6 protein bands from 8 to 260 kDa. Use for easy visualization of gel migration and protein size, and to orient your gel or membrane quickly.
Catalog No. NC0769376 Supplier LICORbio Supplier No. 92880000
The Chameleon 800 Pre-stained Protein Ladder offers multi-colored, pre-stained proteins for both visible and 800 channel near-infrared detection. It includes 6 protein bands from 8 to 260 kDa. Use for easy visualization of gel migration and protein size, and to orient your gel or membrane quickly.