Learn More
LICOR Chameleon Duo Pre-stained Protein Ladder (500 L)
Supplier:  LICOR 92860000
Encompass_Preferred
The Chameleon Duo Pre-stained Protein Ladder provides multi-colored, pre-stained bands for visual inspection and two-color near-infrared detection. It includes 11 protein bands from 8 to 260 kDa. Use the ladder for easy visualization of gel migration and protein size, and to orient your gel or membrane quickly.
Catalog No. NC0738562