Learn More
LICORbio Chameleon Kit Pre-stained Protein Ladders (2 x 250 L)
Catalog No. NC1128447
Encompass_Preferred
The Chameleon Kit provides customizable ladders to suit your experimental needs for near-infrared detection. It includes concentrated solutions of Chameleon 700 and Chameleon 800. Mix and match Chameleon 700 and Chameleon 800 to create the best protein ladder for your experimental needs. It also provides the vibrant, easily visualized qualities of Chameleon Duo.
Catalog No. NC1128447 Supplier LICORbio Supplier No. 92890000
The Chameleon Kit provides customizable ladders to suit your experimental needs for near-infrared detection. It includes concentrated solutions of Chameleon 700 and Chameleon 800. Mix and match Chameleon 700 and Chameleon 800 to create the best protein ladder for your experimental needs. It also provides the vibrant, easily visualized qualities of Chameleon Duo.