Learn More
Grainger Chamfer Profile Router Bit 1-1/2
Catalog No. 502766219
Encompass_Preferred
The Westward Chamfer Profile Router Bit is a 1/4" shank router bit with a 1/2" cutting diameter. It features a 45 degree chamfer profile and is made of high speed steel. This bit is designed for use in handheld or table mounted routers and is ideal for chamfering edges of wood, plastic, and soft metals. The bit has a 1/4" shank diameter and is 1-1/2" long overall. It can be run at speeds up to 24,000 RPM.
Catalog No. 50-276-6219 Supplier Grainger Supplier No. 16Y541
The Westward Chamfer Profile Router Bit is a 1/4" shank router bit with a 1/2" cutting diameter. It features a 45 degree chamfer profile and is made of high speed steel. This bit is designed for use in handheld or table mounted routers and is ideal for chamfering edges of wood, plastic, and soft metals. The bit has a 1/4" shank diameter and is 1-1/2" long overall. It can be run at speeds up to 24,000 RPM.