Learn More
Grainger Chamfer Profile Router Bit 1-1/4
Catalog No. 502765078
Encompass_Preferred
The WESTWARD Chamfer Profile Router Bit is a 1/4" shank router bit with a 1/2" cutting diameter. It features a 45 degree chamfer profile and is made of high speed steel. This bit is designed for use in handheld or table mounted routers and is ideal for chamfering edges of wood, plastic, and soft metals. The bit has a 1/4" shank diameter and is 1-1/2" long overall. It can be run at speeds up to 24,000 RPM. This quality chamfer router bit is made in the USA by WESTWARD, a leading manufacturer of industrial tools and equipment.
Catalog No. 50-276-5078 Supplier Grainger Supplier No. 16Y609
The WESTWARD Chamfer Profile Router Bit is a 1/4" shank router bit with a 1/2" cutting diameter. It features a 45 degree chamfer profile and is made of high speed steel. This bit is designed for use in handheld or table mounted routers and is ideal for chamfering edges of wood, plastic, and soft metals. The bit has a 1/4" shank diameter and is 1-1/2" long overall. It can be run at speeds up to 24,000 RPM. This quality chamfer router bit is made in the USA by WESTWARD, a leading manufacturer of industrial tools and equipment.