Learn More
MSC Hamilton Champion Swivel Caster, 6 in. Diameter, 3000 lbs. Capacity, 1 ea.
Catalog No. 5000512598
Encompass
The Champion Swivel Caster is an industrial-grade caster designed for heavy-duty mobility. Engineered for durability, it features a robust drop-forged steel mounting plate and an integrally forged kingpin design. These casters are protected by a premium powder-coated finish suitable for both interior and exterior use.
  • Capacity of 3,000 lbs.
  • Includes precision tapered thrust bearing for radial thrust control
  • Features CNC-machined 3 3/4 in. diameter main load bearing raceway
  • Equipped with 3/4 in. diameter hex head axle with lock nut
  • Includes ball-check lubrication fittings in swivel assembly and wheel hubs
  • Three year product warranty
Catalog No. 50-005-12598 Supplier MSC Supplier No. 75013110
The Champion Swivel Caster is an industrial-grade caster designed for heavy-duty mobility. Engineered for durability, it features a robust drop-forged steel mounting plate and an integrally forged kingpin design. These casters are protected by a premium powder-coated finish suitable for both interior and exterior use.
  • Capacity of 3,000 lbs.
  • Includes precision tapered thrust bearing for radial thrust control
  • Features CNC-machined 3 3/4 in. diameter main load bearing raceway
  • Equipped with 3/4 in. diameter hex head axle with lock nut
  • Includes ball-check lubrication fittings in swivel assembly and wheel hubs
  • Three year product warranty