Learn More
Electron Microscopy Sciences K-kits Channel Opener
SDP
Catalog No. 501928050
Encompass_Preferred
The Channel Opener is used to remove the channel tips, while the K-kit stays on the carrier. It's made of anodized aluminum with a cut-off slot design at one end. Used with the K-kit for TEM observation of liquid samples.
Catalog No. 50-192-8050 Supplier Electron Microscopy Sciences Supplier No. K7269
The Channel Opener is used to remove the channel tips, while the K-kit stays on the carrier. It's made of anodized aluminum with a cut-off slot design at one end. Used with the K-kit for TEM observation of liquid samples.