Learn More
USA Scientific Inc ChannelMate Reservoir System, 25 mL
Catalog No. NC9895604
Encompass
ChannelMate 25 mL reservoir refill system, 1 reusable polystyrene base and 200 clear polystyrene inserts/pack, sterile
Catalog No. NC9895604 Supplier USA Scientific Inc Supplier No. 13062510
ChannelMate 25 mL reservoir refill system, 1 reusable polystyrene base and 200 clear polystyrene inserts/pack, sterile