Learn More
MSC Chapin Garden Hand Sprayer, 3.5 gal, Chemical Safe, 1 ea
Catalog No. 502869592
Encompass
This 3.5-gallon chemical-safe garden hand sprayer features a coated steel tank, wide mouth, and reinforced hose, making it suitable for concrete applications.
  • Reinforced nylon braid and vinyl hose withstand harsh chemicals
  • Brass nozzle and shut-off with lock-on feature for precise control
  • Chemical-resistant seals prevent leaks and ensure safe operation
  • Poly holster securely holds wand and nozzles
  • Coated steel tank material
  • Volume capacity is 3.5 gal
  • Stream spray pattern
  • Chemical safe design for concrete applications
  • Maximum sprayer pressure of 60 psi
  • Wand length is 24 in
  • Mouth opening diameter is 3 in
Catalog No. 50-286-9592 Supplier MSC Supplier No. 05816590
This 3.5-gallon chemical-safe garden hand sprayer features a coated steel tank, wide mouth, and reinforced hose, making it suitable for concrete applications.
  • Reinforced nylon braid and vinyl hose withstand harsh chemicals
  • Brass nozzle and shut-off with lock-on feature for precise control
  • Chemical-resistant seals prevent leaks and ensure safe operation
  • Poly holster securely holds wand and nozzles
  • Coated steel tank material
  • Volume capacity is 3.5 gal
  • Stream spray pattern
  • Chemical safe design for concrete applications
  • Maximum sprayer pressure of 60 psi
  • Wand length is 24 in
  • Mouth opening diameter is 3 in