Learn More
MSC Chapin Industrial Sprayer, 2 gal, Stainless Steel, Handheld, 1 ea
Catalog No. 502868058
Encompass
This industrial sprayer features a 2-gallon stainless steel tank and is designed for handheld use in various industrial applications. It includes a reinforced hose and a wide mouth opening for easy filling.
  • Wide Tri-Lock opening
  • Brass shut-off with lock-on feature for fatigue-free spraying
  • Handheld design for industrial applications
  • Stainless steel tank
  • Adjustable brass nozzle
  • Reinforced hose, 42 inch length
  • Wand is 12 inch brass
Catalog No. 50-286-8058 Supplier MSC Supplier No. 94268398
This industrial sprayer features a 2-gallon stainless steel tank and is designed for handheld use in various industrial applications. It includes a reinforced hose and a wide mouth opening for easy filling.
  • Wide Tri-Lock opening
  • Brass shut-off with lock-on feature for fatigue-free spraying
  • Handheld design for industrial applications
  • Stainless steel tank
  • Adjustable brass nozzle
  • Reinforced hose, 42 inch length
  • Wand is 12 inch brass