Learn More
MSC Chapin Sprayer Hose, 48 inch, Reinforced Nylon
Catalog No. 502836472
Encompass
This 48-inch reinforced nylon sprayer hose is an ideal replacement or upgrade for various sprayer models. Designed with a filter to prevent debris from entering the tank, it helps maintain the longevity and performance of your equipment. This hose is a genuine replacement component for compatible sprayer systems.
  • 48-inch reinforced nylon construction for durability
  • Includes a filter to prevent debris accumulation
  • Compatible with XP, GP, and backpack sprayers
  • Ensures proper fit and function as a genuine replacement part
Catalog No. 50-283-6472 Supplier MSC Supplier No. 74841966
This 48-inch reinforced nylon sprayer hose is an ideal replacement or upgrade for various sprayer models. Designed with a filter to prevent debris from entering the tank, it helps maintain the longevity and performance of your equipment. This hose is a genuine replacement component for compatible sprayer systems.
  • 48-inch reinforced nylon construction for durability
  • Includes a filter to prevent debris accumulation
  • Compatible with XP, GP, and backpack sprayers
  • Ensures proper fit and function as a genuine replacement part