Learn More
Research Products International Corp CHAPS [3-[(3-Cholamidopropyl)-dimethylammonio]-1-propanesulfonate], 10 Grams
SDP
Catalog No. 50488594
Encompass_Preferred
Zwitterionic, non-denaturing detergent for solubilizing membrane proteins.
Catalog No. 50-488-594 Supplier Research Products International Corp Supplier No. C4101010.0
Zwitterionic, non-denaturing detergent for solubilizing membrane proteins.