Learn More
Selleck Chemical LLC CHAPS
SDP
Supplier:  Selleck Chemical LLC S69971G
Encompass_Preferred
CHAPS is a zwitterionic surfactant that decreases the sequence specificity of the nucleosome
Catalog No. 50-000-02625