Learn More
Selleck Chemical LLC CHAPS
SDP
Catalog No. 5000002414
Encompass_Preferred
CHAPS is a zwitterionic surfactant that decreases the sequence specificity of the nucleosome
Catalog No. 50-000-02414 Supplier Selleck Chemical LLC Supplier No. S6997100MG
CHAPS is a zwitterionic surfactant that decreases the sequence specificity of the nucleosome