Learn More
Gold Biotechnology Inc CHAPS 25 g
SDP
Catalog No. 5000562080
Encompass_Preferred
CHAPS also known as ((3-cholamidopropyl)dimethylammonio)-1-propanesulfonate is a zwitterionic surfactant primarily used to solubilize membrane proteins and other macromolecules
Catalog No. 50-005-62080 Supplier Gold Biotechnology Inc Supplier No. C08025
CHAPS also known as ((3-cholamidopropyl)dimethylammonio)-1-propanesulfonate is a zwitterionic surfactant primarily used to solubilize membrane proteins and other macromolecules