Learn More
Research Products International Corp CHAPS [3-[(3-Cholamidopropyl)-dimethylammonio]-1-propanesulfonate], 25 Grams
SDP
Supplier:  Research Products International Corp C4101025.0
Encompass_Preferred
Zwitterionic, non-denaturing detergent for solubilizing membrane proteins.
Catalog No. 50-213-237