Learn More
Bioworld CHAPS Lysis Buffer (1X), 500 mL
Catalog No. 50253087
Encompass_Preferred
CHAPS Lysis buffer for extracting proteins from cells.Contains:Tris-HCL, pH 7.4 (50mM)NaCl (110mM)EDTA (5mM)Chaps (1%) - CHAPS is 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate
Catalog No. 50-253-087 Supplier Bioworld Supplier No. 220500062
CHAPS Lysis buffer for extracting proteins from cells.Contains:Tris-HCL, pH 7.4 (50mM)NaCl (110mM)EDTA (5mM)Chaps (1%) - CHAPS is 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate