Learn More
Bioworld CHAPS, 100 g
Catalog No. 50255810
Encompass_Preferred
A nondenaturing Zwitterionic detergent suitable for use as a solubilizing agent for membrane proteins due to the disruption of non-specific protein interactions. Ref: Hjelmeland, L.H., Proc. Nat. Acad. Sci. USA, 77, 6368 (1980) Specifications:Ultra Pure GradeConductivity (10%, Water): <=50umhosResidue on Ignition: <=0.1%Purity (TLC): One spot method, >99.9%.Appearance: SolidDensity : 1.01 g/cm3 at 20 °CMelting Point: 157 °C (dec.)
Catalog No. 50-255-810 Supplier Bioworld Supplier No. 403300205
A nondenaturing Zwitterionic detergent suitable for use as a solubilizing agent for membrane proteins due to the disruption of non-specific protein interactions. Ref: Hjelmeland, L.H., Proc. Nat. Acad. Sci. USA, 77, 6368 (1980) Specifications:Ultra Pure GradeConductivity (10%, Water): <=50umhosResidue on Ignition: <=0.1%Purity (TLC): One spot method, >99.9%.Appearance: SolidDensity : 1.01 g/cm3 at 20 °CMelting Point: 157 °C (dec.)