Learn More
Alkali Scientific CHAPS [3 [(3 Cholamidopropyl) dimethylammonio] 1 propanesulfonate], 10 G
SDP
Catalog No. 5000660374
Encompass_Preferred
CHAPS is a zwitterionic detergent widely used in protein solubilization, particularly for membrane proteins. The 10g format provides enough material for larger-scale solubilization and protein studies. CHAPS solubilizes proteins while maintaining their functional structures, making it ideal for applications like chromatography, electrophoresis, and structural analysis. It is less harsh than ionic detergents, offering a mild alternative that preserves protein integrity. CHAPS is commonly used in proteomics, structural biology, and other applications requiring the extraction and study of membrane proteins in their native functional forms.
Catalog No. 50-006-60374 Supplier Alkali Scientific Supplier No. CHA002CP
CHAPS is a zwitterionic detergent widely used in protein solubilization, particularly for membrane proteins. The 10g format provides enough material for larger-scale solubilization and protein studies. CHAPS solubilizes proteins while maintaining their functional structures, making it ideal for applications like chromatography, electrophoresis, and structural analysis. It is less harsh than ionic detergents, offering a mild alternative that preserves protein integrity. CHAPS is commonly used in proteomics, structural biology, and other applications requiring the extraction and study of membrane proteins in their native functional forms.