Learn More
Alkali Scientific CHAPS [3 [(3 Cholamidopropyl) dimethylammonio] 1 propanesulfonate], 100 Grams
SDP
Catalog No. 5000660376
Encompass_Preferred
CHAPS (3-[(3-Cholamidopropyl)dimethylammonio]propane sulfonate) is a zwitterionic detergent used for membrane protein solubilization, offering mild, efficient protein extraction while preserving protein function. This 100g formulation is ideal for applications in membrane protein studies, including purification, structural analysis, and biochemical assays. CHAPS is often preferred in techniques such as electrophoresis, chromatography, and crystallization due to its ability to stabilize proteins in their native state without disrupting their activity. It is frequently used in laboratory experiments requiring the extraction and solubilization of membrane-bound proteins, enzymes, and cellular components.
Catalog No. 50-006-60376 Supplier Alkali Scientific Supplier No. CHA005CP
CHAPS (3-[(3-Cholamidopropyl)dimethylammonio]propane sulfonate) is a zwitterionic detergent used for membrane protein solubilization, offering mild, efficient protein extraction while preserving protein function. This 100g formulation is ideal for applications in membrane protein studies, including purification, structural analysis, and biochemical assays. CHAPS is often preferred in techniques such as electrophoresis, chromatography, and crystallization due to its ability to stabilize proteins in their native state without disrupting their activity. It is frequently used in laboratory experiments requiring the extraction and solubilization of membrane-bound proteins, enzymes, and cellular components.