Learn More
Research Products International Corp CHAPS [3-[(3-Cholamidopropyl)-dimethylammonio]-1-propanesulfonate], 5 Grams
SDP
Supplier:  Research Products International Corp C410105.0
Encompass_Preferred
Zwitterionic, non-denaturing detergent for solubilizing membrane proteins.
Catalog No. 50-213-238