Learn More
Bioworld CHAPS Lysis Buffer (2X), 50 mL
Catalog No. 50253091
Encompass_Preferred
This buffer is used for cell lysis and protein extraction.1x Concentration Contains:50mM Tris-HCl (pH 7.4)110mM NaCl5mM EDTA1% CHAPS.CHAPS is 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate Warning:For laboratory use only.Not fit for agricultural, household, clinical use or humanconsumption.
Catalog No. 50-253-091 Supplier Bioworld Supplier No. 220500082
This buffer is used for cell lysis and protein extraction.1x Concentration Contains:50mM Tris-HCl (pH 7.4)110mM NaCl5mM EDTA1% CHAPS.CHAPS is 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate Warning:For laboratory use only.Not fit for agricultural, household, clinical use or humanconsumption.