Learn More
Biotium CHAPS, Ultra Pure
Catalog No. 501964400
Encompass_Preferred
CHAPS, 3-[(3-cholamidopropyl)dimethylammonio]-1-propanesulfonate, is a zwitterionic detergent that is used for protecting the native state of proteins. This zwitterionic detergent is useful for membrane protein solubilization when it is important to maintain protein activity. It is electrically neutral and easily removed by dialysis.Properties: White solid soluble in water; Store at room temperature; C32H58N2O7S; MW: 614.88; [75621-03-3].
Catalog No. 50-196-4400 Supplier Biotium Supplier No. 2202410G
CHAPS, 3-[(3-cholamidopropyl)dimethylammonio]-1-propanesulfonate, is a zwitterionic detergent that is used for protecting the native state of proteins. This zwitterionic detergent is useful for membrane protein solubilization when it is important to maintain protein activity. It is electrically neutral and easily removed by dialysis.Properties: White solid soluble in water; Store at room temperature; C32H58N2O7S; MW: 614.88; [75621-03-3].