Learn More
Abcam CHAPS, Zwitterionic non-denaturing detergent, 10G
Catalog No. 502215304
Encompass_Preferred
MW 614.9 Da, Purity >98%. Zwitterionic non-denaturing detergent that protects the native state of proteins, able to disrupt non-specific protein interactions and has less protein aggregation than non-ionic detergents. It is electrically neutral and easily removed by dialysis. Often used in the solubilization and purification of membrane proteins.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5304 Supplier Abcam Supplier No. AB14139610G
MW 614.9 Da, Purity >98%. Zwitterionic non-denaturing detergent that protects the native state of proteins, able to disrupt non-specific protein interactions and has less protein aggregation than non-ionic detergents. It is electrically neutral and easily removed by dialysis. Often used in the solubilization and purification of membrane proteins.
The product is subject to the following: Abcam Restricted Use Statement