Learn More
Bioworld CHAPSO, 10 g
Catalog No. 50255813
Encompass_Preferred
CHAPSO3- [(3-Cholamidopropyl) dimethylammonio]-2-hydroxy-1-propanesulfonate] A nondenaturing zwitterionic detergent suitable for use as a solubilizing agent for membrane proteins Ref: MacDonald, R.C., Biochim. Biophys. Acta, 733, 210 (1983) SpecificationsUltra Pure GradeAppearance: SolidMelting Point: 184-186 °C(lit.)
Catalog No. 50-255-813 Supplier Bioworld Supplier No. 403300283
CHAPSO3- [(3-Cholamidopropyl) dimethylammonio]-2-hydroxy-1-propanesulfonate] A nondenaturing zwitterionic detergent suitable for use as a solubilizing agent for membrane proteins Ref: MacDonald, R.C., Biochim. Biophys. Acta, 733, 210 (1983) SpecificationsUltra Pure GradeAppearance: SolidMelting Point: 184-186 °C(lit.)