Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CHAPSO (3-[(3-cholamidopropyl)dimethylammonio]-2-hydroxy-1-propanesulfonate)

Catalog No. p-200019982
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERA65501 1 g
FERA65502 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERA65501 Supplier Thermo Scientific™ Supplier No. A65501
Only null left
Add to Cart
Edge
Add to Cart

CHAPSO, or 3-[(3-cholamidopropyl)dimethylammonio]-2-hydroxy-1-propanesulfonate, is a zwitterionic detergent commonly used in biochemical and biophysical research.

CHAPSO, or 3-[(3-cholamidopropyl)dimethylammonio]-2-hydroxy-1-propanesulfonate, is a zwitterionic detergent commonly used in biochemical and biophysical research. It is widely employed for membrane protein solubilization, stabilization, and purification. CHAPSO is known for its mildness and ability to preserve the structural integrity and functionality of membrane proteins during extraction. Its unique properties make it suitable for a variety of applications, including protein crystallization, spectroscopy, and structural biology studies. With its versatile and effective solubilization capabilities, CHAPSO is a valuable tool for researchers working with membrane proteins.

Features of CHAPSO include:
• Zwitterionic detergent
• Gentle—enabling preservation of native conformation and activity
• Effectively disrupts lipid-protein interactions
• Exhibits low critical micelle concentration (CMC)
• High-purity compound with low UV absorptivity

Properties
CMC: (H2O) ~8 mM (0.50%)
Aggregation number: (H2O) ~11
Purity: ≥99% (by HPLC analysis)
pH: 5–8 (1% solution in water)
Solubility: ≥0.5 M (in water at 20 ̊C)
Fluorescence: <10% (0.1% solution in water at 345 nm)
Absorbance of a 1% solution in water: 260 nm: <0.06, 280 nm: <0.04, 340 nm: <0.02

TRUSTED_SUSTAINABILITY

Specifications

Color White
Product Type Detergent
CAS 82473-24-3
Molecular Formula C32H58N2O8S
Solubility ≥0.5 M (in water at 20°C)
Content And Storage Store below -20°C.
Form Lyophilized
Reagent Type Detergent
Purification Target Proteins
Quantity 1 g
Molecular Weight (g/mol) 630.9
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.