Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Charcoal Powder
SCI_ED

Catalog No. S25243 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
250 g
500 g
Packaging:
Activated Charcoal
Bone Ash
Charcoal Powder
Decolorizing Agent
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25243 500 g Bone Ash
S25244 500 g Charcoal Powder
S25245 250 g Decolorizing Agent
S25246 500 g Activated Charcoal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. S25243
Add to Cart
Add to Cart
This item is not returnable. View return policy

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.011
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Color Black
Density 1.8 to 2.1g/mL
Packaging Bone Ash
Linear Formula C
Quantity 500 g
Formula Weight 12.01
Chemical Name or Material Charcoal Powder
Safety handling Check Alert storage code Red
ShelfLife 5 years
DOTInformation DOT Class 4.2, III, : Spontaneously Combustible
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.