Learn More
LICOR Charcoal Canisters (pk of 8)
Supplier:  LICOR 9300820
Encompass_Preferred
Use these charcoal canisters with the Pearl Small Animal Imaging System for in vivo imaging experiments. The canister removes isofluorane gas from the exhaust air stream.
Catalog No. NC2225418