Learn More
Electron Microscopy Sciences Charcoal Cartridge
SDP
Catalog No. 5025512
Encompass_Preferred
Absorb Incubator Contaminants This disposable, activated charcoal cartridge efficiently absorbs low energy 35S radioactive components during metabolic labeling. The containment problem exists in incubators and is caused by 35S-labeled amino acids, such as methionine and cysteine which are used to label proteins. The charcoal is supplied in a 100 mm diameter plastic petri dish with a highly porous membrane sealed to the top edge.
Catalog No. 50-255-12 Supplier Electron Microscopy Sciences Supplier No. 6445405
Absorb Incubator Contaminants This disposable, activated charcoal cartridge efficiently absorbs low energy 35S radioactive components during metabolic labeling. The containment problem exists in incubators and is caused by 35S-labeled amino acids, such as methionine and cysteine which are used to label proteins. The charcoal is supplied in a 100 mm diameter plastic petri dish with a highly porous membrane sealed to the top edge.