Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Charcoal, Dextran Coated

Catalog No. C62415G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
20 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C62415G 5 g
50-311-9134 20 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C62415G Supplier Sigma Aldrich Supplier No. C62415G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Charcoal, Dextran Coated is made from acid washed charcoal powder and dextran, molecular weight 64-76kDa, combined in a 10:1 (w/w) ratio.Charcoal, Dextran Coated has been used:as a supplement in cell culture media to test the functioning of androgen receptor in the procedure of melatonin radioimmunoassay for the preparation of steroid-free human serumto reduce the creatinine level in untreated serum.The use of dextran-coated charcoal makes the immunoassay of insulin in biological fluids simpler and more rapid. In theory, the charcoal coated with dextran will adsorb the free hormone and leave hormones that are bound to a carrier (or antibody). DextranDry preparation of dextran-treated charcoal

Specifications

Molecular Formula C6H8O7 -+ H2O
Quantity 5 g
MDL Number MFCD00149972
Molecular Weight (g/mol) 210.14 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.